(±)-1,2-Propanediol ≥99.5% (by GC) USP, FCC, Macron Fine Chemicals™
Catalog # 6263-19
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pack type:Steel drum, phenolic lined
- Pk:20 L
- Seq:5302
Specifications
Specification Test Results
- Bulk Pharmaceutical Chemical:= 99.5 %
- USP - Assay (CH₃CHOHCH₂OH) (by GC):≥ 99.5 %
- USP - Identificaton A (IR):Passes Test
- USP - Identification B - Diethylene Glycol:≤ 0.10 %
- USP - Identification B - Ethylene Glycol:≤ 0.10 %
- USP - Identification C:Passes Test
- USP - Acidity:Passes Test
- USP - Water (H₂O)(by Karl Fischer titrn):≤ 0.2 %
- USP - Residue on Ignition:≤ 0.007 %
- USP - Chloride (Cl):≤ 0.007 %
- USP - Sulfate (SO₄):≤ 0.006 %
- FCC - Assay (CH₃CHOHCH₂OH) (by GC):≥ 99.5 %
- FCC - Acidity:Passes Test
- FCC - Identificaton A (IR):Passes Test
- FCC - Lead (Pb):≤ 1 mg/kg
- FCC - Residue on Ignition:≤ 0.007 %
- FCC - Specific Gravity at 25°/25°C:1.035 - 1.037
- FCC - Water (H₂O)(by Karl Fischer titrn):≤ 0.20 %
- FCC - Distilling Range(Lower):185 - 189 °C
- FCC - Distilling Range(Upper):185 - 189 °C
Specification Test Results
About this item
:
:76.1 g/mol
:Management of Change (MOC) category = R
Flash Pt:99 °C
Storage Temperature:Ambient
CAS Number:57-55-6
Formula:CH₃CH(OH)CH₂OH
MDL Number:MFCD00064272