Potassium permanganate ≥99.0%, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 3228-07
Supplier: Avantor
CAS Number:
Specifications
- Pk:12 kg
- Seq:4187
- Environmentally Preferable:
Specifications
Specification Test Results
- Meets ACS Reagent Chemical Requirements:—
- For Laboratory,Research,or Manufacturing Use:= 99.0 %
- Assay (KMnO₄):≥99.0 %
- Insoluble Matter:≤0.2 %
- Chloride and Chlorate (as Cl):≤0.005 %
- Sulfate (SO₄):≤0.02 %
Specification Test Results
About this item
Synonyms: Permanganic acid potassium salt
:
:158.03 g/mol
UN:1490
ADR:5.1,II
:GHS03
:GHS07
:GHS09
CAS Number:7722-64-7
Formula:KMnO₄
MDL Number:MFCD00011364