Potassium carbonate sesquihydrate 98.5-101.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
: Avantor
:6381-79-9
:
:CK₂O₃·1,5H₂O
:MFCD00149925
:165.23 g/mol
:891 °C
:2,29 g/cm³ (20 °C)
:Management of Change (MOC) category = N
:209-529-3
:GHS07
:Ambient
- ACS - Assay (K₂CO₃·1.5H₂O):98.5 - 101.0 %
- ACS - Insoluble Matter:≤0.01 %
- ACS - Loss on Heating at 285°C:14.0 - 16.5 %
- ACS - Calcium (Ca):≤0.005 %
- ACS - Chloride (Cl):≤0.003 %
- ACS - Magnesium (Mg):≤0.002 %
- ACS - Nitrogen Compounds (as N):≤0.001 %
- ACS - Phosphate (PO₄):≤0.001 %
- ACS - Silica (SiO₂):≤0.005 %
- ACS - Sulfur Compounds (as SO₄):≤0.004 %
- ACS - Ammonium Hydroxide Precipitate:≤0.01 %
- ACS - Sodium (Na):≤0.02 %
- Trace Impurities - ACS - Heavy Metals (as Pb):≤5 ppm
- Trace Impurities - Iron (Fe):≤5 ppm
Frequently Bought Together