Order Entry
India
ContactUsLinkComponent
 
L-Threonine 98.5-101.5% (dried basis) USP, Multi-Compendial, J.T.Baker®
Catalog # 2089-06
Supplier:  Avantor
CAS Number:  
undefined
L-Threonine 98.5-101.5% (dried basis) USP, Multi-Compendial, J.T.Baker®
Catalog # 2089-06
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Poly bottle, white, blue T/E cap
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Food GMP Manufactured Product:
    98.5 - 101.5%
  • GMP Manufactured Product:
    Passes Test
  • Meets B.P. Chemical Specifications:
    -29.1 - -26.7 Degree
  • Meets E.P. Chemical Specifications:
    5.0 - 6.5
  • Meets J.P. Chemical Specifications:
    ≤0.2%
  • Meets U.S.P Requirements:
    ≤0.4%
  • Preserve in well-closed containers.:
    ≤0.05%
  • Store in well-closed,light-resistant containers:
    ≤0.03%
  • CAUTION: For Manufacturing, processing or repackaging:
    ≤0.003%
  • Bulk Food Chemical:
    ≤0.5%
  • Bulk Pharmaceutical Chemical:
    ≤2.0%
  • USP - Assay (C₄H₈NO₃) (dried basis):
    98.5 - 101.5 %
  • USP - Identification:
    Passes Test
  • USP - Specific Rotation α²⁵D:
    -29.1 - -26.7 Degree
  • USP - pH:
    5.0 - 6.5
  • USP - Loss on Drying at 105°C:
    ≤ 0.2 %
  • USP - Residue on Ignition:
    ≤ 0.4 %
  • USP - Chloride (Cl):
    ≤ 0.05 %
  • USP - Sulfate (SO₄):
    ≤ 0.03 %
  • USP - Iron (Fe):
    ≤ 0.003 %
  • USP - Related Compounds - Individual Impurities:
    ≤ 0.5 %
  • USP - Related Compounds - Total Impurities:
    ≤ 2.0 %
  • EP/BP - Assay (C₄H₈NO₃) (dried basis):
    99.0 - 101.0 %
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - pH:
    5.0 - 6.5
  • EP/BP - Specific Rotationα²ºD:
    -29.0 - -27.6 Degree
  • EP/BP-Ninhydrin-Positive Substances-Each:
    ≤ 0.2 %
  • EP/BP-Ninhydrin-Positive Substances-Total Impurities:
    ≤ 0.5 %
  • EP/BP - Chloride (Cl):
    ≤ 200 ppm
  • EP/BP - Sulfate (SO₄):
    ≤ 300 ppm
  • EP/BP - Ammonium (NH₄):
    ≤ 0.02 %
  • EP/BP - Iron (Fe):
    ≤ 10 ppm
  • EP/BP - Loss on Drying:
    ≤ 0.5 %
  • EP/BP - Ash (sulfated):
    ≤ 0.1 %
  • JP - Assay (C₄H₈NO₃) (dried basis):
    98.5 - 101.0 %
  • JP - Identification:
    Passes Test
  • JP - Optical Rotation:
    -29.0 - -26.0 Degree
  • JP - pH:
    5.2 - 6.2
  • JP - Clarity and Color of Solution:
    Passes Test
  • JP - Chloride (Cl):
    ≤ 0.021 %
  • JP - Sulfate (SO₄):
    ≤ 0.028 %
  • JP - Ammonium (NH₄):
    ≤ 0.02 %
  • JP - Heavy Metals (as Pb):
    ≤ 20 ppm
  • JP - Arsenic (As):
    ≤ 2 ppm
  • JP - Related Substances:
    Passes Test
  • JP - Loss on Drying at 105°C:
    ≤ 0.20 %
  • JP - Residue on Ignition:
    ≤ 0.10 %

Specification Test Results

About this item

 :
 :119.12 g/mol
Storage Temperature:Ambient
EINECS:200-774-1
CAS Number:72-19-5
Formula:C₄H₉NO₃
MDL Number:MFCD00064270