
Guanidinium hydrochloride, Biotech Reagent, J.T.Baker®
Catalog # 0510-F9
Supplier: Avantor
CAS Number:
Specifications
- Pk:50 kg
- Seq:4418
Specifications
Specification Test Results
- Assay (NH₂C(:NH)NH₂·HCl):99.5 - 101.0 %
- Acidity (as HCl):≤0.01 %
- Loss on Drying at 105°C:≤0.5 %
- Residue after Ignition:≤0.050 %
- Trace Impurities - Copper (Cu):≤0.200 ppm
- Heavy Metals (as Pb):≤0.5 ppm
- Trace Impurities - Iron (Fe):≤2.0 ppm
- Enzyme Activity - DNase Activity:Passes Test
- Enzyme Activity - RNase Activity:Passes Test
- Enzyme Activity - Protease Activity:Passes Test
- Absorbance of a 6M Aqueous Solution - 230 nm:≤0.200
- Absorbance of a 6M Aqueous Solution - 260 nm:≤0.030
- Absorbance of a 6M Aqueous Solution - 275 nm:≤0.030
- Arsenic and Antimony (as As):≤0.500 ppm
- Nitrate (NO₃):≤0.005 %
- Sulfate (SO₄):≤0.005 %
- Cyanide (CN):≤0.1 ppm
- Particulate Matter:Passes Test
- Water-Insoluble Matter:≤0.3 %
- Melamine and Related Compounds:Passes Test
Specification Test Results
About this item
:
:95.53 g/mol
:Management of Change (MOC) category = R
:GHS07
CAS Number:50-01-1
Formula:NH₂C(=NH)NH₂·HCl
MDL Number:MFCD00013026