


Iron(II) sulphate, dried ≥97.0% (Fe basis), fine powder USP, FCC, Macron Fine Chemicals™
Catalog # 5051-30
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pack type:Bag
- Pk:50 kg
- Seq:4418
Specifications
Specification Test Results
- Meets F.C.C. Requirements:Passes Test
- Meets U.S.P Requirements:Passes Test
- Food GMP Manufactured Product:≤3 ppm
- GMP Manufactured Product:≤2 mg/kg
- CAUTION: For Manufacturing, processing or repackaging:≤1 mg/kg
- Bulk Pharmaceutical Chemical:86.0 - 89.0%
- Bulk Food Chemical:Passes Test
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Arsenic (As):≤3 ppm
- USP - Lead (Pb):≤2 mg/kg
- USP - Mercury (Hg):≤1 mg/kg
- USP - Assay (as FeSO4):86.0 - 89.0 %
- FCC - Identification A:Passes Test
- FCC - Identification B:Passes Test
- FCC - Insoluble Substances:≤0.05 %
- FCC - Lead (Pb):≤2 mg/kg
- FCC - Mercury (Hg):≤1 mg/kg
- FCC - Assay (as FeSO4):86.0 - 89.0 %
- Mesh - Thru U.S. No. 40 Sieve:≥97.0 %
- Mesh - Thru U.S. No. 100 Sieve:≥85.0 %
Specification Test Results
About this item
:
:151.91 g/mol
:GHS09
:GHS07
:GHS08
CAS Number:7720-78-7
Formula:FeO₄S
MDL Number:MFCD00011014