Order Entry
India
ContactUsLinkComponent
1-Chlorobutane, ChromAR® for HPLC, Macron Fine Chemicals™
1-Chlorobutane, ChromAR® for HPLC, Macron Fine Chemicals™
Catalog # 6725-25
CAS Number:  
undefined
1-Chlorobutane, ChromAR® for HPLC, Macron Fine Chemicals™
Catalog # 6725-25
CAS Number:  

Specifications

  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

About this item

 :
 :92.57 g/mol
UN:1127
ADR:3,II
CAS Number:109-69-3
Formula:CH₃(CH₂)₃Cl
MDL Number:MFCD00001009