Order Entry
India
ContactUsLinkComponent
 
J.T.Baker® BAKER INSTRA-ANALYZED®, Tin, atomic absorption spectrocopy (AAS) Standard
  6471-04
 :  Avantor
 :  
undefined
J.T.Baker® BAKER INSTRA-ANALYZED®, Tin, atomic absorption spectrocopy (AAS) Standard
  6471-04
 :  Avantor
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    20% HCl
  • Pk:
    150 mL
  • Element:
    Tin
  • Symbol:
    Sn

 

 

Tin standard solution for AAS are prepared from metals and salts of 99,999% spectral purity.

 : Traceable to NIST and SRM numbers.
 : Standard is packaged in a poly bottle.