Order Entry
India
ContactUsLinkComponent
J.T.Baker® BAKER INSTRA-ANALYZED®, Bismuth Standard Solution, 1000 µg/L Bi in Water, Reagent for AAS
J.T.Baker® BAKER INSTRA-ANALYZED®, Bismuth Standard Solution, 1000 µg/L Bi in Water, Reagent for AAS
Catalog # 6445-04
Supplier:  Avantor
CAS Number:  
undefined
J.T.Baker® BAKER INSTRA-ANALYZED®, Bismuth Standard Solution, 1000 µg/L Bi in Water, Reagent for AAS
Catalog # 6445-04
Supplier:  Avantor
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    150 mL
  • Element:
    Bismuth
  • Symbol:
    Bi

Specifications

About this item

Single element atomic absorption standards are used for quantitative analysis and for standardisation of instruments used in a variety of elemental analysis applications.

Certifications: Traceable to NIST and SRM.