J.T.Baker® BAKER INSTRA-ANALYZED®, Iron Standard Solution, 1,000 µg/l Fe, reagent for AAS
Catalog # 6453-04
Supplier: Avantor
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:5% HNO₃
- Pack type:Plastic bottle
- Pk:150 mL
- Element:Iron
- Symbol:Fe
Specifications
About this item
Iron standard solution for AAS are prepared from metals and salts of 99,999% spectral purity.
Certifications: Traceable to NIST and SRM numbers.