Order Entry
India
ContactUsLinkComponent
J.T.Baker® BAKER INSTRA-ANALYZED®, Copper Standard Solution, 1000 µg/l Cu, Reagent for AAS
J.T.Baker® BAKER INSTRA-ANALYZED®, Copper Standard Solution, 1000 µg/l Cu, Reagent for AAS
Catalog # 6451-04
Supplier:  Avantor
CAS Number:  
undefined
J.T.Baker® BAKER INSTRA-ANALYZED®, Copper Standard Solution, 1000 µg/l Cu, Reagent for AAS
Catalog # 6451-04
Supplier:  Avantor
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    150 mL
  • Element:
    Copper
  • Symbol:
    Cu

Specifications

About this item

Copper standard solution for AAS are prepared from metals and salts of 99,999% spectral purity.

Certifications: Traceable to NIST and SRM numbers.