Order Entry
India
ContactUsLinkComponent
Patent blue V calcium salt ≥85% (by titrimetric analysis)
Patent blue V calcium salt ≥85% (by titrimetric analysis)
Catalog # A8187.0025
Supplier:  PanReac AppliChem
CAS Number:  
Patent blue V calcium salt ≥85% (by titrimetric analysis)
Catalog # A8187.0025
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Assay (titr.):
    Min. 85% 
  • In H₂O insoluble matter:
    Max. 0,20% 
  • Unsulphonated primary aromatic amine :
    Max. 0,01% 
  • Heavy metals (as Pb):
    Max. 0,004% 
  • Subsidiary dyes:
    Max. 2,0% 
  • Arsenic (As):
    Max. 0,0003% 
  • Cadmium (Cd):
    Max. 0,0001% 
  • Mercury (Hg):
    Max. 0,0001% 

Specification Test Results

About this item

 :
 :1159.45 g/mol
Storage Temperature:Ambient
CAS Number:3536-49-0
Formula:C₅₄H₆₂CaN₄O₁₄S₄
MDL Number:MFCD00038746