Manufactured from optically clear virgin polystyrene.
- Printed with lot numbers for ease in traceability
- 100% integrity tested
- Sterilised by gamma irradiation and certified non pyrogenic
- Canted models have a skirt to prevent tipping
Non-treated: Flasks are designed for use in applications where cell attachment is not desired.
TC treated: For optimal cell attachment and growth.
Ultra low attachment surface: Features a covalently bound hydrogel layer that effectively inhibits cellular attachment and minimizes protein absorption, enzyme activation, and cellular activation. Surface is non-cytotoxic, biologically inert, and nondegradable.
CellBIND® surface treatment: improves cell attachment by incorporating more oxygen into the cell culture surface, rendering it more hydrophilic and increasing surface stability. CellBIND® surfaces are nonbiological and require no special handling or storage.
The original plug-seal polyethylene cap can be used in both open and closed systems. Phenolic-style polyethylene caps are designed for open gas exchange applications. The lot number is printed on all flasks for traceability.
- Catalog No:
- 734-1718
- 734-4139
- 734-1214
- 734-1726
- 734-0092
- 734-0094
- 734-2272
- 734-0091
- 734-2274
- 734-1733
- 734-1712
- 734-2275
- 734-1706
- 734-1725
- 734-0093
- 734-1724
- 734-1723
- 734-1722
- 734-0090
- 734-1700
- 734-1717
- 734-2707
- 734-2706
- 734-2705
- 734-1719
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta