Order Entry
India
ContactUsLinkComponent
VWR®, Hygiene Masks, Maximum Protection
  VWRU414004-667
undefined
VWR®, Hygiene Masks, Maximum Protection
  VWRU414004-667
  • Type:
    With ear loops
  • Colour:
    Blue
  • Seq:
    01
  • Environmentally Preferable:
  • Pk:
    500
VWR® Maximum Protection face masks feature a three-ply construction for superior particle and bacterial filtration efficiency. Available in two styles, with anti-fog protection or moisture resistance.
  • Latex-free
  • Guards against bacteria and particulates
  • Highly breathable
  • Soft and hypoallergenic
Anti-fog masks: To help prevent protective eyewear from fogging, a soft 20 mm foam strip is positioned at the top of each mask to collect moisture. Spandex ear loops are ultrasonically welded to maintain softness and protect against particulates. A full-length plastic arch within the mask's protective layers holds the fabric away from the wearer's mouth to allow cooler, more comfortable breathing without restricting downward vision. An encased 12 cm malleable steel noseband creates a secure facial seal and improves wearer comfort.
Moisture-resistant cleanroom masks: A lightweight inner film layer provides enhanched moisture resistance. Spandex earloops are ultrasonically welded to maintain softness and protect against particulates. An encased 12 cm malleable steel noseband creates a secure facial seal and improves wearer comfort.
 : Masks are rigorously tested and manufactured in an ISO Certified facility under stringent process controls to ensure that each product meets exacting quality standards. Products are validated through independent laboratory testing.
 : 50 masks per pack, 10 packs per case.