Order Entry
India
Orders LinkContactUsLinkComponent
Baculovirus expression system, flashBAC™
Supplier:   MIRUS
Baculovirus expression system, flashBAC™
Supplier:  MIRUS
Certificates
Baculovirus expression system, flashBAC™
4 Options Available Below
Product Details & Documents
The flashBAC™ Baculovirus Expression System is designed for high yield protein production in insect and mammalian cells.
  • Single-step baculovirus generation in insect cells, no plaque purification required
  • Ready to use with most transfer vectors
  • High recombinant protein yields
This system is a streamlined platform for the production of recombinant baculoviruses. Genetic optimisation of the Autographa californica nucleopolyhedrovirus (AcMNPV) genome yields recombinant virus in a single step and eliminates the need for separation of recombinant and parental virus by plaque purification.
Product Family Options
Product Information
  • Pk
    Availability
    Price
  • MIRUMIR6115
    flashBAC™ Baculovirus expression system, 5 reactions
    flashBAC™ Baculovirus expression system, 5 reactions
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT
  • MIRUMIR6120
    flashBAC™ Baculovirus expression system, 24 reactions
    flashBAC™ Baculovirus expression system, 24 reactions
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT
  • MIRUMIR6135
    flashBAC™ ULTRA baculovirus expression system, 5 reactions
    flashBAC™ ULTRA baculovirus expression system, 5 reactions
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT
  • MIRUMIR6140
    flashBAC™ ULTRA baculovirus expression system, 24 reactions
    flashBAC™ ULTRA baculovirus expression system, 24 reactions
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT