Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
Rs.0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
India
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Penicillin/Streptomycin/Amphotericin B
Penicillin/Streptomycin/Amphotericin B 100X
Penicillin/Streptomycin/Amphotericin B 100X
Penicillin/Streptomycin/Amphotericin B 100X
Catalog # MIRUMIR5970
Supplier:
MIRUS
CAS Number:
Expand image
Penicillin/Streptomycin/Amphotericin B 100X
Catalog # MIRUMIR5970
Supplier:
MIRUS
CAS Number:
Specifications
Pk:
1 KIT
Seq:
0075
Pk Info:
1x100ml
Specifications
About this item
Product Details
:
Product Details