Order Entry
India
Orders LinkContactUsLinkComponent
Transfection kit and reagent, TransIT-PRO®

Transfection kit and reagent, TransIT-PRO®

Supplier:   MIRUS
Transfection kit and reagent, TransIT-PRO®
Supplier:  MIRUS
Certificates
Transfection kit and reagent, TransIT-PRO®

4 Options Available Below

Product Details & Documents

TransIT-PRO® transfection kit and reagents are ideal for biotherapeutic protein production in suspension CHO and 293 cells.

  • High protein yield in suspension CHO and 293 cells, including CHO-S and Expi293F™ cells
  • Compatible with multiple CHO media formulations; the same media to be used for both transient and stable expression
  • Decreases time to produce usable protein by maximising target protein yields through transient transfection
  • Simple workflow enables reproducible protein expression with minimal optimisation

Delivery information: Kit contains TransIT-PRO® transfection reagent and PRO boost reagent. TransIT-PRO® transfection reagent also available separately.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • MIRUMIR5700
  • MIRUMIR5740
  • MIRUMIR5750
  • MIRUMIR5760
  • Cell specific:
  • 293, CHO
  • 293, CHO
  • 293, CHO
  • 293, CHO
  • Bioproduction:
  • Yes
  • Yes
  • Yes
  • Yes
  • DNA:
  • Yes
  • Yes
  • Yes
  • Yes
  • Virus production:
  • Yes
  • Yes
  • Yes

Specifications

Product Family Options

Product Information

  • Pk
    Availability
    Price
  • MIRUMIR5700
    TransIT-PRO® transfection kit, 1,0 ml, with 1,5 ml PRO boost reagent
    TransIT-PRO® transfection kit, 1,0 ml, with 1,5 ml PRO boost reagent
    1 mL
    Log in for availability
    Log in for availability
    1 * 1 mL
    1 * 1 mL

    Specifications:
    More
    Specifications
    Cat. No.
    MIRUMIR5700
    Cell specific
    293, CHO
    Bioproduction
    Yes
    DNA
    Yes
    Virus production
    Yes
  • MIRUMIR5740
    TransIT-PRO® transfection reagent, 1×1,0 ml
    TransIT-PRO® transfection reagent, 1×1,0 ml
    1 mL
    Log in for availability
    Log in for availability
    1 * 1 mL
    1 * 1 mL

    Specifications:
    More
    Specifications
    Cat. No.
    MIRUMIR5740
    Cell specific
    293, CHO
    Bioproduction
    Yes
    DNA
    Yes
    Virus production
  • MIRUMIR5750
    TransIT-PRO® transfection reagent, 1×10 ml
    TransIT-PRO® transfection reagent, 1×10 ml
    10 mL
    Log in for availability
    Log in for availability
    1 * 10 mL
    1 * 10 mL

    Specifications:
    More
    Specifications
    Cat. No.
    MIRUMIR5750
    Cell specific
    293, CHO
    Bioproduction
    Yes
    DNA
    Yes
    Virus production
    Yes
  • MIRUMIR5760
    TransIT-PRO® transfection kit, 10 ml, with 15 ml PRO boost reagent
    TransIT-PRO® transfection kit, 10 ml, with 15 ml PRO boost reagent
    10 mL
    Log in for availability
    Log in for availability
    1 * 10 mL
    1 * 10 mL

    Specifications:
    More
    Specifications
    Cat. No.
    MIRUMIR5760
    Cell specific
    293, CHO
    Bioproduction
    Yes
    DNA
    Yes
    Virus production
    Yes