Transfection reagent, TransIT®-Keratinocyte
Supplier: MIRUS
Certificates
About this item
TransIT®-Keratinocyte transfection reagent is specifically optimised to provide exceptional transfection efficiency of plasmid DNA in primary and immortalised keratinocytes.
- High transfection efficiency (20 to 30%)
- Suitable for both transient and stable transfection
- Low cellular toxicity: Maintain cell density and reduce experimental bias
Transfection with TransIT®-Keratinocyte reagent does not require medium changes and can be carried out in serum-containing medium.
Specifications
- Cell specific:Keratinocyte
- DNA:Yes