Transfection reagent, TransIT®-293
Supplier: MIRUS
Certificates
About this item
TransIT®-293 transfection reagent is specifically optimised to provide exceptional transfection efficiency of plasmid DNA in HEK 293 and cell types of associated lineage.
- High transfection efficiency (75 to 85%)
- Deliver single or multiple plasmids – ideal for specialised applications, such as virus production
- Suitable for both transient and stable transfection
- Low cellular toxicity: Maintain cell density and reduce experimental bias
Transfections using TransIT®-293 reagent do not require medium changes and can be carried out in serum-containing medium. TransIT®-293 can be used for many applications, such as gene expression, viral production, shRNA expression and promoter analysis.
Specifications
- Cell specific:293
- DNA:Yes
- Virus production:Yes