Order Entry
India
ContactUsLinkComponent
Cleanroom wipes, Kimtech™ Pure W5
Cleanroom wipes, Kimtech™ Pure W5
Catalog # KIMB06179
Supplier:  KIMBERLY CLARK
Cleanroom wipes, Kimtech™ Pure W5
Catalog # KIMB06179
Supplier:  KIMBERLY CLARK

Specifications

  • Packed:
    100 wipes/bag
  • Colour:
    White
  • L×W:
    229×229 mm
  • Seq:
    01
  • Model:
    W5
  • Pk:
    100

Specifications

About this item

These critical task wipes feature a 50% rayon/50% polyester spunlace blend base sheet with high water absorbency rate, plus excellent absorption efficiency and capacity.

  • No binders or surfactants are used resulting in low extractables
  • Clothlike feel
  • Double bagged with case liner

Suitable for use in Class 100, ISO 5, M2.5 or higher cleanrooms.

Colour: white

Packaging: Double-bagged in anti-stat packaging.