
(±)-α-Lipoic acid, high purity
Catalog # 0494-25G
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass, amber
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Identification (IR):PASS
- Purity:≥ 99.0 %
- Solubility (5%, EtOH):PASS
Specification Test Results
About this item
:
:206.33 g/mol
Boiling Pt:160 – 165 °C
Melting Pt:59…62 °C
:GHS07
Storage Temperature:Ambient
CAS Number:1077-28-7
Formula:C₈H₁₄O₂S₂
MDL Number:MFCD00005474