Order Entry
India
ContactUsLinkComponent
Filter holder
Filter holder
Catalog # HACH1352900
Supplier:  HACH
undefined
Filter holder
Catalog # HACH1352900
Supplier:  HACH

Specifications

  • For Ø:
    47 mm
  • Description:
    Filter holder, magnetic
  • Seq:
    01
  • Environmentally Preferable:
  • Pk:
    1

Specifications

About this item

This filter holder is made of rugged, autoclavable polyphenylsulfone.

  • Magnetic with no-leak seal
  • One-hand operation for filter retrieval
Delivery: Supplied with support screen and graduated funnel 300 ml (50 ml increments). Stem fits #8 stopper (not included).