Order Entry
India
Orders LinkContactUsLinkComponent
VWR® RNA Loading Buffer (2X)
VWR® RNA Loading Buffer (2X)
RNA loading buffer with ethidium bromide, 2X, 1 mL
Catalog # 1B1607-1ML
Supplier:   VWR Chemicals
VWR® RNA Loading Buffer (2X)
Catalog # 1B1607-1ML
Supplier:  VWR Chemicals
CAS Number:  
VWR® RNA Loading Buffer (2X)
VWR® RNA Loading Buffer (2X)
RNA loading buffer with ethidium bromide, 2X, 1 mL
CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 mL
Product Details & Documents
RNA loading buffers (2X) are premixed loading dyes for the preparation of RNA samples for agarose or polyacrylamide electrophoresis.
  • With ethidium bromide, 2X provides immediate post-electrophoresis visualization of the RNA
  • Efficient RNA separation
  • Contains formamide as a denaturing agent and RNA stabilizer, as well as bromophenol blue and xylene cyanol FF tracking dyes for monitoring of RNA migration
Documents
Specifications
Specifications
  • Pk:
    1 mL
  • Description:
    RNA loading buffer with ethidium bromide, 2X
Specifications