Bovine serum albumin (BSA)
: BIOWEST
Bovine serum albumin (BSA) is commonly used in cell culture protocols, particularly where protein supplementation is necessary and the other components of serum are unwanted. In cell culture, its main role is as a carrier of small molecules. Because of its negative charge, BSA binds water, salts, fatty acids, vitamins and hormones, then carries these bound components between tissues and cells. The binding capacity also makes BSA an effective scavenger to remove toxic substances, including pyrogens, from the medium.
- Source:Serum
- Species:Bovine
- Form:lyophilised
- Protein synonyms:BSA|Fraction V|Cohn Fraction V
- Protein/peptide name:Bovine Serum Albumin
- pH:7
Frequently Bought Together