Order Entry
India
ContactUsLinkComponent
VWR® Budapest, Safety Spectacles
VWR® Budapest, Safety Spectacles
Catalog # 111-1847
Supplier:  VWR Collection
undefined
VWR® Budapest, Safety Spectacles
Catalog # 111-1847
Supplier:  VWR Collection
Specifications
  • Type:
    Anti-UV (2-1,2), anti-scratch, anti-fog
  • Lenses:
    Clear PC
  • Pk:
    1
Specifications
About this item
Wrap-around protective eyewear combining fashion, fit and function. One-piece PC lens and a lightweight, frameless design ensures a comfortable custom fit all day long.
  • Adjustable temple length enables wearers to fit the eyewear to their facial profile
  • Pantoscopic adjustment allows lens angle modification
  • Integrated side shield
  • Soft nose bridge for extra comfort
Certifications: Compliant with EN 166, EN 170; VWR® EN 166 FT CE (frame); VWR® 2C-1,2 1 FT (lens)