VWR® Toronto, Safety Goggles
111-1840
- Type:Anti-scratch, anti-fog
- Colour:Translucent blue
- Lenses:Clear PC
- Pk:1
Wrap-around modern style provides excellent panoramic vision, high level of comfort and maximum protection.
- PC lens, PVC frame, polyester/rubber headband
- Fitted with indirect ventilation
- Opalescent frame prevents light reflection
- Designed to give optimum fit to all facial contours
- Comfortably fits over most spectacles
: Compliant with: EN 166:2001. Frame: EN166 3 4 9 BT CE. Lens: VWR® 1 BT 9.