Disposable samplers, MultiDispo
: BURKLE
This MultiDispo zone samplers for taking samples from multiple layers of the sample material.
- Easy emptying through the opening (Ø 14 mm) in the handle
- Clean room manufactured
- Cross contamination is completely eliminated
Suitable for free-flowing powders and small-grain granulates.
- Catalog No:
- 300-0248
- 300-0245
- 300-0246
- 300-0247
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001 Certificate
- ISO14001 Certificate
- ISO 14001 Certificate
- ISO 14001 Certificate
Frequently Bought Together