Volatile acid standards
705-2015
: HACH
:
- Concentration:62500 mg/l as acetic acid
- Pk:16
- Description:Volatile acid standard solution, pack of 16×10 ml voluette ampoules
Acetate standard solution for accuracy checks in volatile acids determinations.