Order Entry
India
ContactUsLinkComponent
Potassium standard solution
Potassium standard solution
Catalog # 705-2020
Supplier:  HACH
CAS Number:  
undefined
Potassium standard solution
Catalog # 705-2020
Supplier:  HACH
CAS Number:  

Specifications

  • Concentration:
    250 mg/l as K
  • Pk:
    16
  • Description:
    Potassium standard solution, pack of 16×10 ml voluette ampoules (NIST)

Specifications

About this item

Standard solution for accuracy checks in potassium determinations.