Ferroin (1,10-phenanthrolineferrous sulphate complex) (7 mg/ml FeSO₄) in aqueous solution indicator, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Certificates
About this item
CAS Number:14634-91-4
:
Formula:C₃₆H₂₄FeN₆SO₄
MDL Number:MFCD00036428
:692.54 g/mol
EINECS:238-676-6
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:Passes Test
- Suitability:Passes Test
Specification Test Results
Frequently Bought Together