2,6-Dichlorophenolindophenol sodium salt, J.T.Baker®
Catalog # H116-01
Supplier: Avantor
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Loss on Drying at 120°C:= 12.0 %
- Interfering Dyes:Passes Test
Specification Test Results
About this item
:
:290.08 g/mol
Merck Index:13,03093
Storage Temperature:Ambient
EINECS:210-640-4
CAS Number:620-45-1
Formula:C₁₂H₆Cl₂NNaO₂