Dodecamolybdophosphoric acid hydrate, crystals, BAKER ANALYZED® ACS, J.T.Baker®
: Avantor
:51429-74-4
:
:H₃[P(Mo₃O₁₀)₄]·nH₂O
:MFCD00149913
:1843.27 g/mol
:12,07498
:78…90 °C
:1,62 g/cm³ (25 °C)
:Ambient
:234-713-5
:3260
:8,III
:GHS05
:GHS07
- Meets ACS Reagent Chemical Requirements:—
- For Laboratory,Research,or Manufacturing Use:—
- Insoluble Matter:≤ 0.01 %
- Chloride (Cl):≤ 0.02 %
- Sulfate (SO₄):≤ 0.025 %
- Ammonium (NH₄):≤ 0.01 %
- Calcium (Ca):≤ 0.02 %
- Heavy Metals (as Pb):≤ 0.005 %
- Iron (Fe):≤ 0.005 %
Frequently Bought Together