
meta-Phosphoric acid stabilised, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:37267-86-0
:
Formula:(HPO₃)n
MDL Number:MFCD00011351
:79.98 g/mol
Boiling Pt:260 °C (1013 hPa)
Melting Pt:21 °C
Density:2,4 g/cm³ (20 °C)
EINECS:253-433-4
ADR:8,III
:Stabilised with sodium metaphosphate
UN:3260
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Meets ACS Reagent Chemical Requirements:33.5 - 36.5 %
- For Laboratory,Research,or Manufacturing Use:57.0 - 63.0 %
- ACS - Assay (HPO₃) (by acidimetry):33.5 - 36.5 %
- ACS - Stabilizer (as NaPO₃):57.0 - 63.0 %
- ACS - Nitrate (NO₃):≤ 0.001 %
- ACS - Sulfate (SO₄):≤ 0.005 %
- ACS - Chloride (Cl):≤ 0.001 %
- Trace Impurities - Arsenic (As):≤ 1 ppm
- Heavy Metals (as Pb):≤ 0.005 %
- Trace Impurities - Iron (Fe):≤ 0.005 %
- Substances Reducing Permanganate (as H₃PO₃):≤ 0.02 %
Specification Test Results
Frequently Bought Together