Order Entry
India
ContactUsLinkComponent
Aluminium sulphate hexadecahydrate 98.0-102.0%, crystals, AR® ACS, Macron Fine Chemicals™
Corrosive
Health
Environment
Aluminium sulphate hexadecahydrate 98.0-102.0%, crystals, AR® ACS, Macron Fine Chemicals™
Certificates
undefined
Corrosive
Health
Environment
Aluminium sulphate hexadecahydrate 98.0-102.0%, crystals, AR® ACS, Macron Fine Chemicals™
Certificates

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
  • For Laboratory,Research,or Manufacturing Use:
  • ACS - Assay (Al₂(SO₄)₃·16H₂O):
    98.0 - 102.0 %
  • ACS - Calcium (Ca):
    ≤0.01 %
  • ACS - Chloride (Cl):
    ≤0.005 %
  • ACS - Heavy Metals (as Pb):
    ≤0.001 %
  • ACS - Insoluble Matter:
    ≤0.01 %
  • ACS - Iron (Fe):
    ≤0.002 %
  • ACS - Magnesium (Mg):
    ≤0.002 %
  • ACS - Potassium (K):
    ≤0.005 %
  • ACS - Sodium (Na):
    ≤0.02 %
  • Solubility (5 in 100):
    Passes Test
  • ACS - Water (H₂O)(by Karl Fischer titrn), For Information Only:

Specification Test Results

Related Information

Frequently Bought Together