Order Entry
India
ContactUsLinkComponent
 
Sodium hydrogen carbonate 99.7-100.3% (dried basis), powder, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 3506-01
Supplier:  Avantor
CAS Number:  
undefined
Sodium hydrogen carbonate 99.7-100.3% (dried basis), powder, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 3506-01
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Poly bottle, white, blue T/E cap
  • Pk:
    500 g
  • Seq:
    3645
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
    99.7 - 100.3%
  • For Laboratory,Research,or Manufacturing Use:
    ≤0.015%
  • Assay (NaHCO₃) (dried basis):
    99.7 - 100.3 %
  • Insoluble Matter:
    ≤0.015 %
  • Chloride (Cl):
    ≤0.003 %
  • Phosphate (PO₄):
    ≤0.001 %
  • Sulfur Compounds (as SO₄):
    ≤0.003 %
  • Calcium (Ca):
    ≤0.02 %
  • Trace Impurities - Iron (Fe):
    ≤0.001 %
  • Magnesium (Mg):
    ≤0.005 %
  • Potassium (K):
    ≤0.005 %
  • Ammonium (NH₄):
    ≤5 ppm
  • Trace Impurities - ACS - Heavy Metals (as Pb):
    ≤5 ppm

Specification Test Results

About this item

Synonyms: Sodium bicarbonate
 :
 :84.01 g/mol
Melting Pt:270
Density:2.20 g/cm³
Storage Temperature:Ambient
EINECS:205-633-8
CAS Number:144-55-8
Formula:NaHCO₃
MDL Number:MFCD00003528
Merck Index:13,08655