
Lithium carbonate ≥99% (by acidimetry), BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 2362-01
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Poly bottle
- Pk:500 g
- Seq:3645
Specifications
Specification Test Results
- Assay (Li₂CO₃) (by acidimetry):≥99.0 %
- Insoluble in Dilute HCl:≤0.01 %
- Chloride (Cl):≤0.005 %
- Total Sulfur (as SO₄):≤0.2 %
- Heavy Metals (as Pb):≤0.002 %
- Iron (Fe):≤0.002 %
- Calcium (Ca):≤0.01 %
- Potassium (K):≤0.005 %
- Sodium (Na):≤0.010 %
- Nitrate (NO₃):≤5 ppm
Specification Test Results
About this item
:
:73.89 g/mol
Merck Index:13,05549
Melting Pt:720 °C
Density:2,11 g/cm³ (20 °C)
:Management of Change (MOC) category = N
EINECS:209-062-5
:GHS07
Storage Temperature:Ambient
CAS Number:554-13-2
Formula:Li₂CO₃
MDL Number:MFCD00011084