Potassium bromate ≥99.8% (by iodometry), BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 2992-01
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 g
- Seq:3645
Specifications
Specification Test Results
- Assay (KBrO₃) (by iodometry):≥99.8 %
- Insoluble Matter:≤0.005 %
- pH of 5% Solution at 25°C:5.0 - 9.0
- Bromide (Br):Passes Test
- Sulfate (SO₄):≤0.005 %
- Iron (Fe):≤0.002 %
- Sodium (Na):≤0.010 %
- Trace Impurities - ACS - Heavy Metals (as Pb):≤5 ppm
Specification Test Results
About this item
:
:167 g/mol
Merck Index:12,07779
Melting Pt:350 °C
Density:3,218 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-829-8
UN:1484
ADR:5.1,II
:GHS03
:GHS06
:GHS08
CAS Number:7758-01-2
Formula:KBrO₃
MDL Number:MFCD00011359