Order Entry
India
ContactUsLinkComponent
di-Sodium tetraborate decahydrate 99.5-100.5%, BAKER ANALYZED® ACS, J.T.Baker®
Health
di-Sodium tetraborate decahydrate 99.5-100.5%, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 3570-05
Supplier:  Avantor
CAS Number:  
undefined
Health
di-Sodium tetraborate decahydrate 99.5-100.5%, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 3570-05
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Poly bottle
  • Pk:
    2,5 kg
  • Seq:
    4076

Specifications

Specification Test Results

  • ACS - Assay (Na₂B₄O₇·10H₂O):
    99.5 - 105.0 %
  • ACS - Insoluble Matter:
    ≤0.005 %
  • ACS - pH of 0.01M Solution at 25°C:
    9.15 - 9.20
  • ACS - Chloride (Cl):
    ≤0.001 %
  • ACS - Phosphate (PO₄):
    ≤0.001 %
  • ACS - Sulfate (SO₄):
    ≤0.005 %
  • ACS - Calcium (Ca):
    ≤0.005 %
  • ACS - Heavy Metals (as Pb):
    ≤0.001 %
  • Trace Impurities - Iron (Fe):
    ≤5 ppm

Specification Test Results

About this item

 :
 :381.37 g/mol
Merck Index:13,08662
Boiling Pt:320 °C (1013 hPa)
Melting Pt:75 °C
Density:1,73 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-540-4
REACH:01-2119490790-32
 :GHS08
CAS Number:1303-96-4
Formula:Na₂B₄O₇·10H₂O
MDL Number:MFCD00149193