


1,4-Dioxane ≥99%, AR® ACS, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Certificates
About this item
CAS Number:123-91-1
:
Formula:C₄H₈O₂
MDL Number:MFCD00006571
:88.11 g/mol
Merck Index:13,03330
Boiling Pt:101,1 °C (1013 hPa)
Melting Pt:11,8 °C
Density:1,03 g/cm³ (20 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:204-661-8
UN:1165
ADR:3,II
:GHS02
:GHS08
:GHS07
Specification Test Results
- Meets ACS Reagent Chemical Requirements:= 99.0 %
- For Laboratory,Research,or Manufacturing Use:= 0.01 %
- ACS - Assay (C₄H₈O₂) (by GC, corrected for water):≥99.0 %
- ACS - Carbonyl Compounds (as HCHO) (by polarography):≤0.01 %
- ACS - Color (APHA):≤20
- ACS - Freezing Point:≥11.0 °C
- ACS - Peroxide (as H₂O₂):≤0.005 %
- ACS - Residue after Evaporation:≤0.005 %
- ACS - Solubility in H₂O:Passes Test
- ACS - Titrable Acid (meq/g):≤0.0016
- ACS - Water (H₂O)(by coulometry):≤0.05 %
Specification Test Results
Frequently Bought Together