Order Entry
India
ContactUsLinkComponent
Sulphanilic acid, anhydrous 98.0-102.0%, powder, BAKER ANALYZED® ACS, J.T.Baker®
 
Sulphanilic acid, anhydrous 98.0-102.0%, powder, BAKER ANALYZED® ACS, J.T.Baker®
  0354-01
 :  Avantor
 :  
undefined
 
Sulphanilic acid, anhydrous 98.0-102.0%, powder, BAKER ANALYZED® ACS, J.T.Baker®
  0354-01
 :  Avantor
 :  

 

  • Pack type:
    Poly bottle, white, blue T/E cap
  • Pk:
    500 g
  • Seq:
    3645

 

 

  • Meets ACS Reagent Chemical Requirements:
    98.0 - 102.0%
  • For Laboratory,Research,or Manufacturing Use:
    ≤0.010%
  • Assay (4-NH₂C₆H₄SO₃H):
    98.0 - 102.0 %
  • Residue after Ignition:
    ≤ 0.010 %
  • Insoluble in Na₂CO₃ Solution:
    ≤ 0.01 %
  • Chloride (Cl):
    ≤ 0.002 %
  • Sulfate (SO₄):
    ≤ 0.005 %
  • Suitability for Nitrogen Dioxide Determination (ASTM D-1607):
    Passes Test
  • Nitrite (NO₂):
    ≤ 0.5 ppm

 

 

 :
 :173.19 g/mol
 :13,09012
 :363 °C (1013 hPa)
 :279 °C
 :1,485 g/cm³ (25 °C)
 :Ambient
 :204-482-5
 :GHS07
 :121-57-3
 :4-(H₂N)C₆H₄SO₃H
 :MFCD00007886