


4-Methyl-2-pentanone ≥98.5% (by GC, corrected for water content), AR® ACS, Macron Fine Chemicals™
:108-10-1
:
:(CH₃)₂CHCH₂COCH₃
:MFCD00008938
:100.16 g/mol
:13,05227
:117,4 °C (1013 hPa)
:–84 °C
:0,801 g/cm³ (20 °C)
:16 °C
:Ambient
:203-550-1
:1245
:3,II
:GHS02
:GHS07
:GHS08
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
- Meets ACS Reagent Chemical Requirements:—
- For Laboratory,Research,or Manufacturing Use:—
- ACS - Assay (CH₃COCH₂CH(CH₃)₂) (by GC, corrected for water):≥98.5 %
- ACS - Color (APHA):≤15
- ACS - Residue after Evaporation:≤0.005 %
- ACS - Titrable Acid (meq/g):≤0.002
- ACS - Water (H₂O):≤0.1 %
- Appearance (clear, colorless liquid):—
Frequently Bought Together