
Diammonium hydrogen citrate 98.0-103.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
: Avantor
:3012-65-5
:
:HOC(CO₂H)(CH₂CO₂NH₄)₂
:MFCD00013068
:226.19 g/mol
:13,00513
:1,48 g/cm³ (20 °C)
:221-146-3
:GHS07
:Ambient
- Meets ACS Reagent Chemical Requirements:98.0 - 103.0%
- For Laboratory,Research,or Manufacturing Use:≤0.005%
- Assay ((NH₄)₂HC₆H₅O₇) (by formol method):98.0 - 103.0 %
- Insoluble Matter:≤0.005 %
- Residue after Ignition:≤0.010 %
- Chloride (Cl):≤0.001 %
- Oxalate (C₂O₄):Passes Test
- Sulfur Compounds (as SO₄):≤0.005 %
- Trace Impurities - Iron (Fe):≤0.001 %
- ACS - Phosphate (PO₄):≤5 ppm
- ACS - Heavy Metals (as Pb):≤5 ppm
Frequently Bought Together