L(+)-Glutamic acid ≥99.0% (dried basis, T), BAKER ANALYZED®, Biochemical Reagent, J.T.Baker®
Catalog # M756-07
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Poly bottle, white, blue T/E cap
- Pk:500 g
- Seq:3645
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:= 99.0 %
- Assay (C₅H₉NO₄) (dried basis, by acid-base titrn):≥99.0 %
- Specific Rotation, [Å]²º^D(dried basis, c=2 in 5N HCl)(+):29.9 - 32.1 Degree
- Homogeneity by TLC:Passes Test
- Ash (sulfated):≤0.1 %
- Loss on Drying at 85°C:≤0.1 %
- Chloride (Cl):≤0.2 %
- Arsenic (As):≤0.0003 %
- Heavy Metals (as Pb):≤0.002 %
- Iron (Fe):≤0.003 %
Specification Test Results
About this item
:
:147.13 g/mol
Merck Index:13,04482
Boiling Pt:325 °C (1013 hPa)
Melting Pt:160 °C
Density:1,538 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-293-7
CAS Number:56-86-0
Formula:HO₂CCH₂CH₂CH(NH₂)CO₂H
MDL Number:MFCD00002634