Order Entry
India
ContactUsLinkComponent
Diethanolamine ≥98.5% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®
Corrosive
Health
Irritant
Diethanolamine ≥98.5% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Corrosive
Health
Irritant
Diethanolamine ≥98.5% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
    = 98.5 %
  • For Laboratory,Research,or Manufacturing Use:
    104.0 - 106.0
  • ACS - Assay ((HOCH₂CH₂)₂NH) (by GC, corrected for water):
    ≥98.5 %
  • ACS - Apparent Equivalent Weight:
    104.0 - 106.0
  • ACS - Color (APHA):
    ≤15
  • ACS - Monoethanolamine:
    ≤1.0 %
  • ACS - Triethanolamine:
    ≤1.0 %
  • ACS - Residue after Ignition:
    ≤0.005 %
  • ACS - Heavy Metals (as Pb):
    ≤0.002 %
  • ACS - Iron (Fe):
    ≤0.001 %
  • ACS - Water (H₂O)(by Karl Fischer titrn):
    ≤0.15 %
  • Chloride (Cl):
    ≤5 ppm

Specification Test Results

Related Information

Frequently Bought Together