



n-Hexane ≥99.0% (by GC), ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
Catalog # N168-08
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle, amber, narrow mouth
- Pk:4 L
- Seq:5162
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:≥99.5%
- ACS - Assay (Total Saturated C₆ Isomers) (by GC, corrected for water):≥99.5 %
- ACS - Assay (as n-Hexane) (by GC, corrected for water):≥99.0 %
- ACS - Color (APHA):≤10
- ACS - Residue after Evaporation:≤3 ppm
- ACS - Water-Soluble Titrable Acid, meq/g:≤0.0003
- ACS - Sulfur Compounds (as S):≤0.005 %
- ACS - Thiophene:Passes Test
- ACS - Water (by KF, coulometric):≤0.03 %
- Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission:≤0.3 ppb
- Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities:≤1.0 ppb
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 400-280 nm:≤0.01
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 250 nm:≤0.02
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 240 nm:≤0.04
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 230 nm:≤0.10
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 220 nm:≤0.20
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 210 nm:≤0.30
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 195 nm:≤1.00
Specification Test Results
About this item
:
:86.18 g/mol
Merck Index:13,04712
Boiling Pt:69 °C (1013 hPa)
Melting Pt:–94,3 °C
Density:0,66 g/cm³ (20 °C)
Flash Pt:–22 °C
Storage Temperature:Ambient
EINECS:203-777-6
UN:1208
ADR:3,II
:Packaged under inert gas.
:GHS02
:GHS08
:GHS07
:GHS09
CAS Number:110-54-3
:92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD00009520