Order Entry
India
ContactUsLinkComponent
2-Propanol ≥99.5%, PHOTREX® for UV spectrophotometry, J.T.Baker®
Flammable
Irritant
2-Propanol ≥99.5%, PHOTREX® for UV spectrophotometry, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Flammable
Irritant
2-Propanol ≥99.5%, PHOTREX® for UV spectrophotometry, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    Min 99.5 %
  • Assay (CH₃CHOHCH₃):
    ≥99.5 %
  • Carbonyl Compounds - Acetone:
    ≤0.002 %
  • Carbonyl Compounds - Propionaldehyde:
    ≤0.002 %
  • Color (APHA):
    ≤10
  • Residue after Evaporation:
    ≤0.0005 %
  • Solubility in H₂O:
    Passes Test
  • ACS - Titrable Acid or Base (µeq/g):
    ≤0.1
  • Water (by KF, coulometric):
    ≤0.05 %
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 400-330 nm:
    ≤0.01
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 300 nm:
    ≤0.02
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 275 nm:
    ≤0.03
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 260 nm:
    ≤0.04
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 245 nm:
    ≤0.08
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 230 nm:
    ≤0.20
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 220 nm:
    ≤0.40
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 210 nm:
    ≤1.00

Specification Test Results

Related Information

Frequently Bought Together