
Citric acid monohydrate 99.0-102.0% (by acidimetry), BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:5949-29-1
:
Formula:HOC(COOH)(CH₂COOH)₂·H₂O
MDL Number:MFCD00149972
:210.14 g/mol
Merck Index:13,02350
Melting Pt:135…152 °C
Density:1,552 g/cm³ (20 °C)
Flash Pt:173,9 °C (closed cup)
EINECS:201-069-1
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Meets ACS Reagent Chemical Requirements:—
- For Laboratory,Research,or Manufacturing Use:—
- Assay (HOC(COOH)(CH₂COOH)₂·H₂O) (by acidimetry):99.0 - 102.0 %
- Insoluble Matter:≤0.005 %
- Residue on Ignition:≤0.02 %
- Chloride (Cl):≤0.001 %
- Oxalate (C₂O₄):Passes Test
- Phosphate (PO₄):≤0.001 %
- Sulfur Compounds (as SO₄):≤0.002 %
- Substances Carbonizable by Hot H₂SO₄:Passes Test
- Trace Impurities - Iron (Fe):≤2.0 ppm
- Trace Impurities - Lead (Pb):≤2.000 ppm
Specification Test Results
Frequently Bought Together