Calcium acetate monohydrate 99.0-101.0%, powder, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:5743-26-0
:MFCD00062576
Formula:(H₃CCOO)₂Ca·1H₂O
MDL Number:MFCD00150019
:176.18 g/mol
Merck Index:13,01645
Density:1,50 g/cm³
Storage Temperature:Ambient
EINECS:200-540-9
Specification Test Results
- Meets ACS Reagent Chemical Requirements:99.0 - 101.0%
- For Laboratory,Research,or Manufacturing Use:Passes Test
- Assay ((CH₃COO)₂CaH₂O):99.0 - 101.0 %
- Alkalinity:Passes Test
- Titrable Acid (meq/g):≤ 0.035
- Insoluble Matter:≤ 0.005 %
- Chloride (Cl):≤ 0.001 %
- Sulfate (SO₄):≤ 0.01 %
- Barium (Ba):≤ 0.005 %
- Heavy Metals (as Pb):≤ 0.001 %
- Trace Impurities - Iron (Fe):≤ 0.001 %
- Nitrate (NO₃):≤ 0.003 %
- Magnesium (Mg):≤ 0.05 %
- Potassium (K):≤ 0.01 %
- Sodium (Na):≤ 0.02 %
- Strontium (Sr):≤ 0.05 %
Specification Test Results
Frequently Bought Together