


Acetic acid glacial ≥99.7% (by GC, corrected for water content), AR® ACS for trace metal analysis, Macron Fine Chemicals™
:64-19-7
:
:H₃CCOOH
:MFCD00036152
:60.05 g/mol
:118 °C (1013 hPa)
:17 °C
:1,05 g/cm³ (20 °C)
:38,5 °C
:Ambient
:200-580-7
:2789
:8,II
:01-2119475328-30
:GHS02
:GHS05
:GHS07
- Meets ACS Reagent Chemical Requirements:Min 99.7 %
- For Laboratory,Research,or Manufacturing Use:Max 0.01 %
- IMPORTANT: Material will freeze if stored below 17 °C (63°F).:Max 0.4 ppm
- ACS - Assay (by GC, corrected for water):≥99.7 %
- ACS - Acetic Anhydride ((CH₃CO)₂O):≤0.01 %
- ACS - Chloride (Cl):≤0.4 ppm
- ACS - Color (APHA):≤10
- Dilution Test:Passes Test
- ACS - Heavy Metals (by ICP-OES):≤5 ppm
- ACS - Residue after Evaporation:≤8 ppm
- ACS - Substances Reducing Dichromate:Passes Test
- ACS - Substances Reducing Permanganate:Passes Test
- ACS - Sulfate (SO₄):≤0.4 ppm
- ACS - Titrable Base (meq/g):≤0.0004
- Trace Impurities - Aluminum (Al):≤0.300 ppm
- Trace Impurities - Arsenic (As):≤0.050 ppm
- Trace Impurities - Boron (B):≤0.200 ppm
- Trace Impurities - Calcium (Ca):≤0.300 ppm
- Trace Impurities - Chromium (Cr):≤0.200 ppm
- Trace Impurities - Copper (Cu):≤0.10 ppm
- Trace Impurities - Gold (Au):≤0.300 ppm
- Trace Impurities - Iron (Fe):≤0.2 ppm
- Trace Impurities - Lead (Pb):≤0.300 ppm
- Trace Impurities - Magnesium (Mg):≤0.300 ppm
- Trace Impurities - Manganese (Mn):≤0.300 ppm
- Trace Impurities - Nickel (Ni):≤0.1 ppm
- Trace Impurities - Potassium (K):≤0.300 ppm
- Trace Impurities - Sodium (Na):≤0.300 ppm
- Trace Impurities - Tin (Sn):≤0.300 ppm
- Trace Impurities - Titanium (Ti):≤0.300 ppm
- Trace Impurities - Zinc (Zn):≤0.300 ppm
Frequently Bought Together