


Acetone ≥99.3% (by GC), BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
: Avantor
:67-64-1
:
:CH₃COCH₃
:MFCD00008765
:58.08 g/mol
:13,00067
:56,2 °C (1013 hPa)
:–95,4 °C
:0,792 g/cm³ (20 °C)
:Ambient
:200-662-2
:1090
:3,II
:Filtered through a 0.2 µm filter, packaged under nitrogen.
:<–20 °C (closed cup)
:01-2119471330-49
:GHS02
:GHS07
:GHS08
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
- For Laboratory, Research,or Manufacturing Use:= 0.01
- Ultraviolet Absorbance (1.00-cm Cell vs. Water) - 400 - 350 nm:≤0.01
- Ultraviolet Absorbance (1.00-cm Cell vs. Water) - UV Cut-off, nm:≤330
- Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission:≤0.3 ppb
- Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities:≤1.0 ppb
- Assay ((CH₃)₂CO) (by GC):≥99.3%
- Residue after Evaporation:≤2.0 ppm
- Titrable Acid (µeq/g):≤0.3
- Titrable Base (µeq/g):≤0.6
- Water (H₂O):≤0.4%
Frequently Bought Together